Order Entry
Denmark
Orders LinkContactUsLinkComponent
Docusate sodium USP

Docusate sodium USP

Fibre drum, 25 kg
Catalog # SPCMDO105-25KGBL
Irritant
Supplier:   Spectrum Chemical
Docusate sodium USP
Catalog # SPCMDO105-25KGBL
Supplier:  Spectrum Chemical
CAS Number:  577-11-7
Docusate sodium USP

Fibre drum, 25 kg

CAS Number:  577-11-7
Price
95.210,00 kr
Log in for availability
1 * 25 kg
Product Details & Documents

Melamine statement (available upon request)
BSE/TSE statement (available upon request)
Allergen statement (available upon request)

Synonyms:  Dioctyl sulphosuccinate sodium salt, Dioctyl sulphosuccinate, sodium salt, Sodium dioctyl sulphosuccinate, Sodium 1,4-bis((2-ethylhexyl)oxy)-1,4-dioxobutane-2-sulphonate, bis(2-Ethylhexyl) sodium sulphosuccinate
  • CAS Number:577-11-7
  • Formula:C₂₀H₃₇NaO₇S
  • MDL Number:MFCD00012455
  • Molecular Weight:444.57 g/mol
  • Merck Index:12,03460
  • Boiling Pt:∼180 °C (1013 hPa)
  • Melting Pt:∼155 °C
  • Density:∼1.1 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:209-406-4

Documents

Specifications

Specification Test Results

  • Assay:
    99% Min.

Specifications

  • Pack type:
    Fibre drum
  • Pk:
    25 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.