Order Entry
Denmark
ContactUsLinkComponent
Kit meter C3210 pH/conductivity/DO complete: C3210 meter, pH electrode SP20T, EC electrode SK20T, 2x50ml pH buffers, 50ml electrolyte, 50ml EC standard, DO electrode SZ20T. 1 * 1 items
Catalog # CONSC3210T
Supplier:  CONSORT
Kit meter C3210 pH/conductivity/DO complete: C3210 meter, pH electrode SP20T, EC electrode SK20T, 2x50ml pH buffers, 50ml electrolyte, 50ml EC standard, DO electrode SZ20T. 1 * 1 items
Catalog # CONSC3210T
Supplier:  CONSORT
Frequently Bought Together