Hydroquinone USP
Glass, amber, 500 g
Catalog # SPCMHY113-500GM
Supplier: Spectrum Chemical
Jump to:
Product Details & Documents
BSE/TSE statement (available upon request)
Allergen statement (available upon request)
Synonyms: 1,4-Dihydroxybenzene, Quinol
- CAS Number:123-31-9
- Formula:C₆H₄(OH)₂
- MDL Number:MFCD00002339
- Molecular Weight:110.11 g/mol
- Merck Index:13,04833
- Boiling Pt:285 °C (1013 hPa)
- Melting Pt:172 °C
- Density:1.332 g/cm³ (20 °C)
- Flash Pt:165 °C
- Storage Temperature:Ambient
- EINECS:204-617-8
- UN:3077
- ADR:9,III
Documents
SPCMHY113-2.5KG_TDS SPCMHY113-500GM_TDS_EN
Product specification
Specifications
Specification Test Results
- Assay:99% Min.
Specifications
- Pack type:Glass, amber
- Pk:500 g



