SCANDIUM STANDARD FOR ICP 1 * 100 mL
Catalog # 92279-100ML
Supplier: Merck
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Syringes, 3-piece
Supplier: TERUMO
VWR®, Nitrile Gloves
Supplier: VWR Collection
Mercury Standard Solution, 1000 mg/L Hg 1 * 100 mL
Supplier: Merck
Tetrahydrofuran ≥99.9% inhibitor-free, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Disodium hydrogen phosphate dihydrate 99+% for HPLC
Supplier: Thermo Fisher Scientific