Order Entry
Denmark
ContactUsLinkComponent
SpectraMax® Time Resolved Fluorescence (TRF) Detection Cartridge is built to measure Europium and Samarium, and is ideal for protein-protein interactions, GPCR and enzyme activity applications. Compatible with SpectraMax® PARADIGM® and/or SpectraMax® 1 * 1 items
Catalog # MLDV0200-7008
Supplier:  Molecular Devices
SpectraMax® Time Resolved Fluorescence (TRF) Detection Cartridge is built to measure Europium and Samarium, and is ideal for protein-protein interactions, GPCR and enzyme activity applications. Compatible with SpectraMax® PARADIGM® and/or SpectraMax® 1 * 1 items
Catalog # MLDV0200-7008
Supplier:  Molecular Devices
Frequently Bought Together