Order Entry
Denmark
Orders LinkContactUsLinkComponent
Affinity chromatography columns, StrepTrap™ XT

Affinity chromatography columns, StrepTrap™ XT

StrepTrap™ XT, PP, 34 µm, 16,00 mm, 25 mm, 5×5 ml, 5 ml/min
Catalog # 29-4013-23
Supplier:   Cytiva
Affinity chromatography columns, StrepTrap™ XT
Catalog # 29-4013-23
Supplier:  Cytiva
Affinity chromatography columns, StrepTrap™ XT
Affinity chromatography columns, StrepTrap™ XT

StrepTrap™ XT, PP, 34 µm, 16,00 mm, 25 mm, 5×5 ml, 5 ml/min

Price
19.070,00 kr
Log in for availability
1 * 5 Items
Product Details & Documents

A chromatography resin for the purification of recombinant proteins tagged with Strep-tag® II and Twin-Strep-tag®, which provides higher affinity, resulting in higher yields and lower loss of valuable target protein.

  • Highly pure Strep-tag® II and Twin-Strep-tag® recombinant proteins
  • Fast purifications with simple elution
  • Physiological conditions and mild elution preserve target protein activity
  • Fast and easy regeneration

Compatible with a wide range of detergents, denaturants, and other additives. Available in 10 and 50 ml lab packs of bulk resin and prepacked in 1 and 5 ml StrepTrap™ XT chromatography columns for convenience.

Specifications
  • Flow rate:
    5 ml/min
  • Ø int.:
    16,00 mm
  • Particle Size:
    34 µm
  • Capacity:
    5×5 ml
  • Description:
    StrepTrap™ XT
  • Length:
    25 mm
  • Seq:
    04
  • Column hardware:
    PP
  • Cat. No.:
    29-4013-23
  • Max. pressure:
    5 bar
  • Storage:
    2...8 °C in 20% ethanol
  • pH range:
    6 - 10
  • Application:
    Purification of recombinant proteins tagged with Strep-tag® II and Twin-Strep-tag®