Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Denmark
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Sodium Hydroxide 0.1M (0.1N). Duree de conservation : 24 mois. Store under normal laboratory conditions, at temperatures between 15° to 25°C 1 * 1 L
Catalog # ACSDZ15171095.1L
Supplier:
CPACHEM
Certificates
Expand image
Sodium Hydroxide 0.1M (0.1N). Duree de conservation : 24 mois. Store under normal laboratory conditions, at temperatures between 15° to 25°C 1 * 1 L
Catalog # ACSDZ15171095.1L
Supplier:
CPACHEM
Certificates
Related Information
Documentation
Certificates
Search by Lot Number
Documentation
Frequently Bought Together
Hydrochloric acid 1 mol/l (1 N), Titripur® Reag. USP, Reag. Ph. Eur. volumetric solution, Supelco®
Supplier: Merck
Test strips, nitrate/nitrite, Dosatest
Supplier: VWR Chemicals
pH indicator papers, MQuant®, Supelco®
Supplier: Merck
Visual colour kits, total hardness, MQuant®, Supelco®
Supplier: Merck
Sodium hydroxide, pellets, EMSURE® for analysis, Supelco®
Supplier: Merck
VWR® Dosatest, pH indicator test strips, non bleeding
Supplier: VWR Chemicals
Ammonia solution 25%, EMSURE® ISO, Reag. Ph. Eur. for analysis, Supelco®
Supplier: Merck
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR® Facial Tissues
Supplier: VWR Collection
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
VWR®, Nitrile Gloves
Supplier: VWR Collection
Test kits, COD cell test, Spectroquant®, Supelco®
Supplier: Merck
Acetonitrile ≥99.9%, LiChrosolv® Reag. Ph. Eur., gradient grade for liquid chromatography, Supelco®
Supplier: Merck
Sodium chloride, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Supplier: Merck
Potassium sulphate ≥99.0%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Test strips for peroxide, MQuant®, Supelco®
Supplier: Merck
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals