TMB ELISA PEROXIDASE SUBSTRATE-FRIENDLY 1 * 500 mL
UPTIUPUP664701
: Uptima
Frequently Bought Together
Safe-Lock Tubes, 2,0 ml - Microtubes
: EPPENDORF
Eppendorf Tubes® 3810X Microtubes 1,5 ml
: EPPENDORF
Eppendorf® Tube Racks
: EPPENDORF
VWR®, Petri Dish, PS
: VWR Collection
VWR® PCR Tubes, 0,2 ml
: VWR Collection
Reagent/centrifuge tube with screw cap
: Greiner Bio-One
VWR® Standard Line Transfer Pipettes
: VWR Collection
Cell culture flasks, Nunc™
: Thermo Fisher Scientific
VWR®, Centrifuge Tubes, Metal-Free, with Screw Cap
: VWR Collection
VWR® Inoculating Loops and Needles, Disposable
: VWR Collection
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
: EPPENDORF
VWR®, Transfer Pipettes, High Performance
: VWR Collection