Order Entry
Denmark
ContactUsLinkComponent
Vivaspin Turbo 15 Regenerated Cellulose membrane, 50,000 MWCO, 12/PK ultrafiltration centrifugal concentrators for protein, DNA, nan oparticle and virus concentration for 4-15 mL sample volumes 1 * 12 items
Catalog # 512-0757
Supplier:  Sartorius
Vivaspin Turbo 15 Regenerated Cellulose membrane, 50,000 MWCO, 12/PK ultrafiltration centrifugal concentrators for protein, DNA, nan oparticle and virus concentration for 4-15 mL sample volumes 1 * 12 items
Catalog # 512-0757
Supplier:  Sartorius
Frequently Bought Together