Order Entry
Denmark
ContactUsLinkComponent
Vivaspin Turbo 15 Regenerated Cellulose membrane, 5,000 MWCO, 48/PK ultrafiltration centrifugal concentrators for protein, DNA, nano particle and virus concentration for 4-15 mL sample volumes 1 * 48 items
Catalog # 512-0752
Supplier:  Sartorius
undefined
Vivaspin Turbo 15 Regenerated Cellulose membrane, 5,000 MWCO, 48/PK ultrafiltration centrifugal concentrators for protein, DNA, nano particle and virus concentration for 4-15 mL sample volumes 1 * 48 items
Catalog # 512-0752
Supplier:  Sartorius
Frequently Bought Together