Order Entry
Denmark
ContactUsLinkComponent
Vivaspin Turbo 15 Regenerated Cellulose membrane, 5,000 MWCO, 12/PK ultrafiltration centrifugal concentrators for protein, DNA, nano particle and virus concentration for 4-15 mL sample volumes 1 * 12 items
Catalog # 512-0751
Supplier:  Sartorius
Vivaspin Turbo 15 Regenerated Cellulose membrane, 5,000 MWCO, 12/PK ultrafiltration centrifugal concentrators for protein, DNA, nano particle and virus concentration for 4-15 mL sample volumes 1 * 12 items
Catalog # 512-0751
Supplier:  Sartorius
Frequently Bought Together