Ammonium Thiocyanate 0.1M (0.1N). Duree de conservation : 12 mois. Store under normal laboratory conditions, at temperatures between 15 to 25C 1 * 1 L
Catalog # ACSDZ12351005.1L
Leverandør: CPACHEM
Ammonium Thiocyanate 0.1M (0.1N). Duree de conservation : 12 mois. Store under normal laboratory conditions, at temperatures between 15 to 25C 1 * 1 L
Catalog # ACSDZ12351005.1L
Leverandør: CPACHEM
Frequently Bought Together
Buffer solutions, ready to use, referenced at 25 °C, Certipur®
Leverandør: Merck
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Leverandør: VWR Chemicals
Titan Yellow Paper Reag. Ph. Eur. 1090901, VWR®
Leverandør: VWR Chemicals
Buffer solutions, Certipur®, colour coded, Supelco®
Leverandør: Merck
pH buffer solutions, certified (DKD)
Leverandør: Mettler - Toledo
pH buffer solutions
Leverandør: Mettler - Toledo
Xylenol orange triturate 1% (w/w) in potassium nitrate Reag. Ph. Eur. 1096301
Leverandør: VWR Chemicals
Pararosaniline solution Reag. Ph. Eur. 1062201, decolourised
Leverandør: VWR Chemicals
BUFFER PH 7,00 DOSING BOTTLE DURACAL 1 * 500 mL
Leverandør: HAMILTON BONADUZ
Buffer solutions in sachets, Certipur®, Supelco®
Leverandør: Merck
pH buffers, DuraCal
Leverandør: HAMILTON BONADUZ
VWR® PESTINORM®, Tetrahydrofuran (THF), Secondary Reference Standard for GC
Leverandør: VWR Chemicals
Potassium chloride 3 mol/l (3 N) in aqueous solution, Supelco®
Leverandør: Merck
pH buffer solutions, colour coded
Leverandør: REAGECON
pH Standard Solutions, IUPAC, Certified
Leverandør: Hach
Reagents for chloride determination using Lovibond® MD photometers
Leverandør: Lovibond
Conductivity standards, Potassium chloride, IUPAC
Leverandør: Hach