Sodium hydroxide solution 10 mol/L EMPROVE EXPERT 1 * 4 L
Catalog # 4.80648.4000
Supplier: MERCK PRODUCTION CHEMICALS
Sodium hydroxide solution 10 mol/L EMPROVE EXPERT 1 * 4 L
Catalog # 4.80648.4000
Supplier: MERCK PRODUCTION CHEMICALS
Frequently Bought Together
Acid Red 183 30% (dye content), Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
ALKALINITY TOTAL PHOTOMETR REAG. 500MG/L 1 * 50 items
Supplier: Palintest
Trisodium 2-(4-Sulphophenylazo)-1,8-dihydroxynaphthalene-3,6-disulphonate (SPADNS) ≥80%, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
NANOCOLOR® Photometric Tube Tests Kits for Ammonium
Supplier: MACHEREY-NAGEL
QUANTOFIX CHLORINE 500 TEST STRIPS 1 * 100 Tests
Supplier: MACHEREY-NAGEL
Bromocresol green-methyl red solution mixed indicator, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Heparin Sodium, Reference Standards
Supplier: USP
Acid red 52, Technical Grade, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Nitrite VHR L, 500 ml
Supplier: Lovibond
Sodium hydroxide 98%, flakes, pure
Supplier: Thermo Fisher Scientific
Reagents for Phosphate Determination using Lovibond® MD photometers
Supplier: Lovibond
Sodium hydroxide, anhydrous ≥98%, pellets, Reagent Grade
Supplier: Honeywell Chemicals