PTFE tapes, ResTape
ResTape, anti-galling, compatible with stainless steel and many other metals and polymers, Grey
REST22487
: Restek
ResTape PTFE tape is for use with threaded connections in a wide range of plumbing materials.
- Each roll is 12,7 mm × 6,6 m
Tapes have a maximum effective temperature of 260 °C.
- Colour:Grey
- Description:ResTape, anti-galling, compatible with stainless steel and many other metals and polymers
- Seq:03



