0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Denmark
ContactUsLinkComponent
Density Standard (20C) UKAS ISO/IEC17025 and ISO 17034 Certified, nominal density 0.7788 G/ML (20C) 1 * 60 mL
PARGDEN20-03
:
PARAGON SCIENTIFIC
Density Standard (20C) UKAS ISO/IEC17025 and ISO 17034 Certified, nominal density 0.7788 G/ML (20C) 1 * 60 mL
PARGDEN20-03
:
PARAGON SCIENTIFIC
Frequently Bought Together
Chloride standard solution certified reference material 1000 mg/l Cl Certipur 1 * 500 mL
: Merck
VWR® 12-Channel Reagent Reservoirs
: VWR Collection
Single wafer shippers, for 4" (100 mm) wafer handling, H 22/H 93 series
: ENTEGRIS