Order Entry
Denmark
Orders LinkContactUsLinkComponent
Condensers for REVOLVE 3M microscope
Condensers for REVOLVE 3M microscope
Swing out universal condenser NA 0,9 WD 0,17 mm
Catalog # ECHLJCND2003
Supplier:   DISCOVER ECHO
Condensers for REVOLVE 3M microscope
Catalog # ECHLJCND2003
Supplier:  DISCOVER ECHO
Condensers for REVOLVE 3M microscope
Swing out universal condenser NA 0,9 WD 0,17 mm
Price
14.310,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Documents
Specifications
  • Description:
    Swing out universal condenser NA 0,9 WD 0,17 mm
  • Seq:
    04