Order Entry
Denmark
ContactUsLinkComponent
Cefotaxime / clavulanic acid disc 30 / 10mcg. Each package should include 50 disc and each Disc should include cefotaxime/clavulanic acid 30 / 10mcg 1 * 250 items
Catalog # HIMESD724-1PK
Supplier:  HIMEDIA
Cefotaxime / clavulanic acid disc 30 / 10mcg. Each package should include 50 disc and each Disc should include cefotaxime/clavulanic acid 30 / 10mcg 1 * 250 items
Catalog # HIMESD724-1PK
Supplier:  HIMEDIA
Frequently Bought Together