Order Entry
Denmark
Orders LinkContactUsLinkComponent
Racks for Axygen® MultiRack tips
Racks for Axygen® MultiRack tips
MultiRack empty rack, 5/pack, 1 pack/case, 10 µl extended length, 200 µl, 200 µl NX, 300 µl tips
Catalog # AXYGMR-R-A
Supplier:   Corning
Racks for Axygen® MultiRack tips
Catalog # AXYGMR-R-A
Supplier:  Corning
Racks for Axygen® MultiRack tips
Racks for Axygen® MultiRack tips
MultiRack empty rack, 5/pack, 1 pack/case, 10 µl extended length, 200 µl, 200 µl NX, 300 µl tips
Price
875,00 kr
Log in for availability
1 * 5 Items
Specifications
  • Description:
    MultiRack empty rack, 5/pack, 1 pack/case
  • Seq:
    01
  • For use with:
    10 µl extended length, 200 µl, 200 µl NX, 300 µl tips