Order Entry
Denmark
ContactUsLinkComponent
ReproSil-Pur Basic-C18, 2.5 μm 10 x 4 mm, guard 1 * 5 items
Catalog # MAISR125.B9.V0104
Supplier:  DR MAISCH
ReproSil-Pur Basic-C18, 2.5 μm 10 x 4 mm, guard 1 * 5 items
Catalog # MAISR125.B9.V0104
Supplier:  DR MAISCH
Frequently Bought Together