Safety faceshields, Shape™
Shape™ safety faceshield, clear visor
Catalog # 111-2594
Supplier: JSP
Jump to:
Product Details & Documents
A compact, comfortable, and lightweight faceshield with superior optical quality.
- Perfect optical quality
- 2,2 mm polycarbonate visor
- Wide field of vision
- Adjustable wheel ratchet headband
- Compatible with prescription glasses
The Shape™ Safety Faceshield features a moulded polycarbonate visor that provides superior optical quality with an exceptional field of vision.
Certifications: EN 166; EN 170.
Documents
TDS Faceshield Shape
Datasheet
Specifications
- Beskrivelse:Shape™ safety faceshield, clear visor
- Seq:02
Anbefalinger