Order Entry
Denmark
ContactUsLinkComponent
VISCOSITY STANDARD, ROTATIONAL MINERAL OIL, UKAS ISO 17025 AND ISO 17034 CERTIFIED, NOMINAL VISCOSITY 1517 MPA.S (25.00C), RTM26 1 * 500 mL
Catalog # PARGRTM26
Supplier:  PARAGON SCIENTIFIC
VISCOSITY STANDARD, ROTATIONAL MINERAL OIL, UKAS ISO 17025 AND ISO 17034 CERTIFIED, NOMINAL VISCOSITY 1517 MPA.S (25.00C), RTM26 1 * 500 mL
Catalog # PARGRTM26
Supplier:  PARAGON SCIENTIFIC
Frequently Bought Together