VISCOSITY STANDARD, ROTATIONAL MINERAL OIL, UKAS ISO 17025 AND ISO 17034 CERTIFIED, NOMINAL VISCOSITY 4.745 MPA.S (25.00C), RTM6 1 * 500 mL
Catalog # PARGRTM6
Supplier: PARAGON SCIENTIFIC
VISCOSITY STANDARD, ROTATIONAL MINERAL OIL, UKAS ISO 17025 AND ISO 17034 CERTIFIED, NOMINAL VISCOSITY 4.745 MPA.S (25.00C), RTM6 1 * 500 mL
Catalog # PARGRTM6
Supplier: PARAGON SCIENTIFIC
Frequently Bought Together
pH buffer solutions
Supplier: Mettler - Toledo
VWR®, Nitrite 1000 mg/L in water standard for ion chromatography
Supplier: VWR Chemicals
Sulphuric acid 1 mol/l (2 N) in aqueous solution Reag. Ph. Eur. 1086804
Supplier: VWR Chemicals
Buffer solutions in sachets
Supplier: Mettler - Toledo
VWR® ARISTAR®, Sodium 1000 mg/L in Water Standard for Ion Chromatography
Supplier: VWR Chemicals
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
VWR®, Sulphate 1000 mg/L in water standard for ion chromatography
Supplier: VWR Chemicals
VWR® Buffer Solutions, Secondary Standards
Supplier: VWR Chemicals