Order Entry
Denmark
ContactUsLinkComponent
VISCOSITY STANDARD, ROTATIONAL MINERAL OIL, UKAS ISO 17025 AND ISO 17034 CERTIFIED, NOMINAL VISCOSITY 3.329 MPA.S (25.00C), RTM5 1 * 500 mL
Catalog # PARGRTM5
Supplier:  PARAGON SCIENTIFIC
VISCOSITY STANDARD, ROTATIONAL MINERAL OIL, UKAS ISO 17025 AND ISO 17034 CERTIFIED, NOMINAL VISCOSITY 3.329 MPA.S (25.00C), RTM5 1 * 500 mL
Catalog # PARGRTM5
Supplier:  PARAGON SCIENTIFIC