VISCOSITY STANDARD, GENERAL PURPOSE, UKAS ISO 17025 AND ISO 17034 CERTIFIED, NOMINAL VISCOSITY 2.144 MPA.S (25 C) 1 * 500 mL
Catalog # PARGN2
Supplier: PARAGON SCIENTIFIC
VISCOSITY STANDARD, GENERAL PURPOSE, UKAS ISO 17025 AND ISO 17034 CERTIFIED, NOMINAL VISCOSITY 2.144 MPA.S (25 C) 1 * 500 mL
Catalog # PARGN2
Supplier: PARAGON SCIENTIFIC
Frequently Bought Together
Phthalate Esters-Mix 1 2000 ng/µl 1 * 1 mL
Supplier: EHRENSTORFER
Buffer Solutions
Supplier: Thermo Fisher Scientific
Isomaltulose, Reference Standard
Supplier: USP
Azimsulfuron certified 1 * 10 mg
Supplier: EHRENSTORFER
Pirimicarb Analytical standard, reference material, Fluka™
Supplier: Honeywell Chemicals
Reagents for chloride determination using Lovibond® MD photometers
Supplier: Lovibond
Orion™ Conductivity Standards and Solutions, Thermo Scientific
Supplier: Thermo Orion
Fonofos certified 1 * 0,1 g
Supplier: EHRENSTORFER
VWR®, Sulphur, Certified Reference Materials, in light mineral oil
Supplier: VWR Chemicals
100mg/l each of As,Be,Bi,Ca,Cd,Co,Cr,Cu,Fe,Li,Mg,Mn,Mo,Ni,Pb,Sb,Se,Sr,Ti,Tl,V,Zn 1 * 500 mL
Supplier: CUSTOM MADE CHEMICALS LAB
Fenoxaprop-P (R-enantiomer) certified 1 * 0,1 g
Supplier: EHRENSTORFER