Order Entry
Denmark
ContactUsLinkComponent
VISCOSITY STANDARD, GENERAL PURPOSE, UKAS ISO 17025 AND ISO 17034 CERTIFIED, NOMINAL VISCOSITY 494 MPA.S (25 °C) 1 * 500 mL
Catalog # PARGD500
Supplier:  PARAGON SCIENTIFIC
VISCOSITY STANDARD, GENERAL PURPOSE, UKAS ISO 17025 AND ISO 17034 CERTIFIED, NOMINAL VISCOSITY 494 MPA.S (25 °C) 1 * 500 mL
Catalog # PARGD500
Supplier:  PARAGON SCIENTIFIC
Frequently Bought Together