Order Entry
Denmark
ContactUsLinkComponent
Custom LCFA Standard in Water Transvaccenic Acid 693-72-1 0.1% Oleic acid 112-80-1 0.1 Stearic acid 57-11-4 0.1 Dodecanoic acid 143-07-7 0.1 Hexadecanoic acid 57-10-3 0.1 Tetradecanoic acid 544-63-8 0.1 Palmitoleic acid 373-49-9 0.1 1 * 5 mL
Catalog # RESTCS-26516
Supplier:  Restek
Custom LCFA Standard in Water Transvaccenic Acid 693-72-1 0.1% Oleic acid 112-80-1 0.1 Stearic acid 57-11-4 0.1 Dodecanoic acid 143-07-7 0.1 Hexadecanoic acid 57-10-3 0.1 Tetradecanoic acid 544-63-8 0.1 Palmitoleic acid 373-49-9 0.1 1 * 5 mL
Catalog # RESTCS-26516
Supplier:  Restek
Frequently Bought Together