Order Entry
Denmark
ContactUsLinkComponent
Dropping point thermometer, similar to Ubbelohde, 0+110:1C, enclosed form, capillary prismatic colourless, red special filling, completely immersed, Richter closure, approx. 240x10mm, state-calibrated, with DKD certificate (DAkkS) with 3 test points 1 * 1 items
Catalog # AMARM235000-FL/DKD
Supplier:  Amarell
Dropping point thermometer, similar to Ubbelohde, 0+110:1C, enclosed form, capillary prismatic colourless, red special filling, completely immersed, Richter closure, approx. 240x10mm, state-calibrated, with DKD certificate (DAkkS) with 3 test points 1 * 1 items
Catalog # AMARM235000-FL/DKD
Supplier:  Amarell
Frequently Bought Together