Order Entry
Denmark
ContactUsLinkComponent
Detect Human B7-1/CD80 with <10pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture sup ernates and serum. This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. 1 * 1 KIT
Catalog # BSBTFEK0707
Supplier:  Boster Bio
Detect Human B7-1/CD80 with <10pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture sup ernates and serum. This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. 1 * 1 KIT
Catalog # BSBTFEK0707
Supplier:  Boster Bio
Frequently Bought Together