Order Entry
Denmark
ContactUsLinkComponent
Detect Rat Fibronectin/FN1 with <15pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates, serum and plasma(heparin, EDTA, citrate). This is a TMB colorimetric sandwich ELISA kit . 1 * 1 KIT
Catalog # BSBTFEK0350
Supplier:  Boster Bio
Detect Rat Fibronectin/FN1 with <15pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates, serum and plasma(heparin, EDTA, citrate). This is a TMB colorimetric sandwich ELISA kit . 1 * 1 KIT
Catalog # BSBTFEK0350
Supplier:  Boster Bio
Frequently Bought Together