Order Entry
Denmark
ContactUsLinkComponent
Detect Human Fas Ligand/FASLG with <2pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell cultu re supernates, serum and plasma(heparin, EDTA, citrate). This is a TMB colorimetric sandwich ELISA kit . 1 * 1 KIT
Catalog # BSBTFEK0337
Supplier:  Boster Bio
Detect Human Fas Ligand/FASLG with <2pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell cultu re supernates, serum and plasma(heparin, EDTA, citrate). This is a TMB colorimetric sandwich ELISA kit . 1 * 1 KIT
Catalog # BSBTFEK0337
Supplier:  Boster Bio
Frequently Bought Together