Order Entry
Denmark
ContactUsLinkComponent
Detect Rat BMP2 with <2pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: bone tissue, cell cultur e supernates and serum. This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up 1 * 1 KIT
Catalog # BSBTFEK0312
Supplier:  Boster Bio
Detect Rat BMP2 with <2pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: bone tissue, cell cultur e supernates and serum. This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up 1 * 1 KIT
Catalog # BSBTFEK0312
Supplier:  Boster Bio
Frequently Bought Together