Order Entry
Denmark
ContactUsLinkComponent
Rabbit IgG polyclonal antibody for BAT3/BAG6 detection. Tested with WB, IHC-P, IHC-F, ICC/IF, FCM, Direct ELISA in Human Mouse Rat. 1 * 100 µG
Catalog # BSBTA00967-1
Supplier:  Boster Bio
Rabbit IgG polyclonal antibody for BAT3/BAG6 detection. Tested with WB, IHC-P, IHC-F, ICC/IF, FCM, Direct ELISA in Human Mouse Rat. 1 * 100 µG
Catalog # BSBTA00967-1
Supplier:  Boster Bio
Frequently Bought Together