31 components 100µg/kg each of Al As B Ba Be Bi Ca Cd Co Cr Cu Fe Ga In K Li Mg Mn Mo Na Ni Pb Se Sr Th Tl U V Y Zn Ag in HNO3 5% 1 * 100 mL
Catalog # ACSDRM030145L1
Supplier: CPACHEM
31 components 100µg/kg each of Al As B Ba Be Bi Ca Cd Co Cr Cu Fe Ga In K Li Mg Mn Mo Na Ni Pb Se Sr Th Tl U V Y Zn Ag in HNO3 5% 1 * 100 mL
Catalog # ACSDRM030145L1
Supplier: CPACHEM
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
VWR® Facial Tissues
Supplier: VWR Collection
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
VWR®, Nitrile Gloves
Supplier: VWR Collection
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Sodium chloride 99.5-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Aluminum, plasma standard solutions, Specpure®
Supplier: Thermo Fisher Scientific
ICP METAL STANDARD 31 COMPONENT 1 * 100 mL
Supplier: CPACHEM