Order Entry
Denmark
ContactUsLinkComponent
The ScanLater Western Blot cartridge option, user-installable to any SpectraMax i3 or SpectraMax Paradigm base system. The ScanLater Western Blot detection system includes the ScanLater Western Blot Detection Cartridge, ScanLater Membrane Holder an 1 * 1 items
Catalog # MLDV0200-7027
Supplier:  Molecular Devices
The ScanLater Western Blot cartridge option, user-installable to any SpectraMax i3 or SpectraMax Paradigm base system. The ScanLater Western Blot detection system includes the ScanLater Western Blot Detection Cartridge, ScanLater Membrane Holder an 1 * 1 items
Catalog # MLDV0200-7027
Supplier:  Molecular Devices
Frequently Bought Together