Order Entry
Denmark
Orders LinkContactUsLinkComponent
Condensers for REVOLVE 3M microscope
Condensers for REVOLVE 3M microscope
Turret condenser hi resol phase NA 0,85 WD 7 mm
Catalog # ECHLJCND2002
Supplier:   DISCOVER ECHO
Condensers for REVOLVE 3M microscope
Catalog # ECHLJCND2002
Supplier:  DISCOVER ECHO
Condensers for REVOLVE 3M microscope
Turret condenser hi resol phase NA 0,85 WD 7 mm
Price
16.700,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Documents
Specifications
  • Description:
    Turret condenser hi resol phase NA 0,85 WD 7 mm
  • Seq:
    05