Order Entry
Denmark
Orders LinkContactUsLinkComponent
Objectives for Rebel Microscopes
Objectives for Rebel Microscopes
Plan apochromat oil, N.A. 1,35, 0,13 mm
Catalog # ECHLRMAG9112
Supplier:   DISCOVER ECHO
Objectives for Rebel Microscopes
Catalog # ECHLRMAG9112
Supplier:  DISCOVER ECHO
Objectives for Rebel Microscopes
Objectives for Rebel Microscopes
Plan apochromat oil, N.A. 1,35, 0,13 mm
Price
27.320,00 kr
Log in for availability
1 * 1 Items
Specifications
  • Magnification:
    100×
  • Description:
    Plan apochromat oil, N.A. 1,35
  • Seq:
    02
  • Working distance:
    0,13 mm
Anbefalinger