Objectives for Rebel Microscopes
Plan achromat phase, N.A. 0,65, 0,4 mm
Catalog # ECHLRMAG0403
Supplier: DISCOVER ECHO
Jump to:
Product Details & Documents
Documents
Specifications
- Magnification:40×
- Description:Plan achromat phase, N.A. 0,65
- Seq:10
- Working distance:0,4 mm
- Cat. No.:ECHLRMAG0403