Order Entry
Denmark
ContactUsLinkComponent
Techspray 1597-8S, Eco-vortex ultra pure duster, with HFO-1234ZE propellant, non-flammable, ultra-low global warming impact, without cfc / hcfc / hfc, odorless, low toxicity, rohs compliant, 12 aerosol cans of 200ml. 1 * 12 items
Catalog # TEXW1597-8S
Supplier:  Texwipe
Techspray 1597-8S, Eco-vortex ultra pure duster, with HFO-1234ZE propellant, non-flammable, ultra-low global warming impact, without cfc / hcfc / hfc, odorless, low toxicity, rohs compliant, 12 aerosol cans of 200ml. 1 * 12 items
Catalog # TEXW1597-8S
Supplier:  Texwipe
Frequently Bought Together