Formamide solution, NMR reference standard, Supelco®
Formamide solution, 90% in DMSO-d6 (99.9 atom % D), NMR reference standard, NMR tube size 5 mm x 8 in., 1
Catalog # 736201-8IN
Supplier: Merck
Formamide solution, NMR reference standard, Supelco®
Catalog # 736201-8IN
Supplier: Merck
CAS Number:
Jump to:
Product Details & Documents
Specifications
Specifications
- Concentration:90% in DMSO-d6 (99.9 atom % D)
- Pk:1
- Standards:NMR reference standard
- Description:Formamide solution
- Pk Info:NMR tube size 5 mm x 8 in.
- Cat. No.:736201-8IN
- Description:Formamide solution
Specifications
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



