Order Entry
Denmark
Orders LinkContactUsLinkComponent
Serological pipettes

Serological pipettes

2 ml, 0,02 ml, +, Green, Individually
Catalog # RATI3102011
Supplier:   RATIOLAB
Serological pipettes
Catalog # RATI3102011
Supplier:  RATIOLAB
Serological pipettes
Serological pipettes

2 ml, 0,02 ml, +, Green, Individually

Price
1.380,00 kr
Log in for availability
1 * 1.000 Items
Product Details & Documents

Disposable pipettes made from clear polystyrene (PS).

  • Free from RNase, DNase, DNA and pyrogens
  • Sterile, individually packed
  • From one mould – hence more accurate and stable and with minimal residual wetting
  • Permanent black embossed graduations
  • Colour coded cotton wool plug for convenient identification
  • Negative graduation for additional capacity
  • 5, 10 and 25 ml pipettes with scales for dispensed and the residual volumes
Specifications
  • Colour code:
    Green
  • Packed:
    Individually
  • Sterile:
    +
  • Capacity:
    2 ml
  • Division:
    0,02 ml
Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.