Order Entry
Denmark
ContactUsLinkComponent
Cable lead-in, Cable lead-in with sleeve nut, components made of PP natural slotted plug of black silicone for tight fastening. Inc luding hexagon wrench and fastening screws. For lead-in cables or hard-walled tubing from OD 0,1 to max. 10 mm. Insta 1 * 1 items
Catalog # BOHLV1782-07
Supplier:  Bohlender
undefined
Cable lead-in, Cable lead-in with sleeve nut, components made of PP natural slotted plug of black silicone for tight fastening. Inc luding hexagon wrench and fastening screws. For lead-in cables or hard-walled tubing from OD 0,1 to max. 10 mm. Insta 1 * 1 items
Catalog # BOHLV1782-07
Supplier:  Bohlender
Frequently Bought Together