Order Entry
Denmark
ContactUsLinkComponent
Glove Box Duo 2 with Transfer Chamber Antistatic, Aluminium frame with static dissipative panels made of polycarbonate, base panel m ade of static dissipative polyethylene. Front and back side with one door each, two pairs of opposite glove ports eac 1 * 1 items
Catalog # BOHLV1715-08
Supplier:  Bohlender
undefined
Glove Box Duo 2 with Transfer Chamber Antistatic, Aluminium frame with static dissipative panels made of polycarbonate, base panel m ade of static dissipative polyethylene. Front and back side with one door each, two pairs of opposite glove ports eac 1 * 1 items
Catalog # BOHLV1715-08
Supplier:  Bohlender
Frequently Bought Together