Order Entry
Denmark
ContactUsLinkComponent
Adaptor Tri-Clamp Hose Connector, Fitting made of PTFE for transition of Tri-Clamp as per DIN 32 676 to a hose connector. Universal chemical resistance, the flowing product is only exposed to PTFE. 1 * 1 items
Catalog # BOHLD915-42
Supplier:  Bohlender
undefined
Adaptor Tri-Clamp Hose Connector, Fitting made of PTFE for transition of Tri-Clamp as per DIN 32 676 to a hose connector. Universal chemical resistance, the flowing product is only exposed to PTFE. 1 * 1 items
Catalog # BOHLD915-42
Supplier:  Bohlender
Frequently Bought Together