Order Entry
Denmark
ContactUsLinkComponent
Tri-Clamp GL-Adaptor, Fitting made of PTFE for transition of Tri-Clamp as per DIN 32 676 to a GL-threaded neck. Universal chemical r esistance, the flowing product is only exposed to PTFE. 1 * 1 items
Catalog # BOHLD901-42
Supplier:  Bohlender
undefined
Tri-Clamp GL-Adaptor, Fitting made of PTFE for transition of Tri-Clamp as per DIN 32 676 to a GL-threaded neck. Universal chemical r esistance, the flowing product is only exposed to PTFE. 1 * 1 items
Catalog # BOHLD901-42
Supplier:  Bohlender
Frequently Bought Together