Order Entry
Denmark
ContactUsLinkComponent
GLS-Reductions, Blue screw cap made of PP, movable reduction body made of PP with O-Ring made of FPM for transition to glthreads. Th e body can be moved independently from the screw cap so that the completely assembled reduction can be removed and fi 1 * 1 items
Catalog # BOHLD787-08
Supplier:  Bohlender
undefined
GLS-Reductions, Blue screw cap made of PP, movable reduction body made of PP with O-Ring made of FPM for transition to glthreads. Th e body can be moved independently from the screw cap so that the completely assembled reduction can be removed and fi 1 * 1 items
Catalog # BOHLD787-08
Supplier:  Bohlender
Frequently Bought Together