Order Entry
Denmark
ContactUsLinkComponent
Propeller Stirrer Shafts EX, PTFE-EX jacketed stainless steel shaft, propeller completely made of PTFE-EX with three 45° angled roun d or angular blades. Good chemical resistance the product is only exposed to PTFE-EX. 1 * 1 items
Catalog # BOHLC278-12
Supplier:  Bohlender
undefined
Propeller Stirrer Shafts EX, PTFE-EX jacketed stainless steel shaft, propeller completely made of PTFE-EX with three 45° angled roun d or angular blades. Good chemical resistance the product is only exposed to PTFE-EX. 1 * 1 items
Catalog # BOHLC278-12
Supplier:  Bohlender
Frequently Bought Together