Order Entry
Denmark
ContactUsLinkComponent
Reactor Lid EX, Made of static dissipative PTFE-EX. Suitable for glass reactors DN 100 with SCHOTT flat flange. With screw-in thread s for connection of transition fittings. Bore with female thread M5x6 for earthing connection on the upper side of th 1 * 1 items
Catalog # BOHLB170-16
Supplier:  Bohlender
undefined
Reactor Lid EX, Made of static dissipative PTFE-EX. Suitable for glass reactors DN 100 with SCHOTT flat flange. With screw-in thread s for connection of transition fittings. Bore with female thread M5x6 for earthing connection on the upper side of th 1 * 1 items
Catalog # BOHLB170-16
Supplier:  Bohlender
Frequently Bought Together