Reactor Lid EX, Made of static dissipative PTFE-EX. Suitable for glass reactors DN 60 with SCHOTT flat flange. With NPT screw-in thr eads for connection of transition fittings. Bore with female thread M5x6 for earthing connection on the upper side of 1 * 1 items
Catalog # BOHLB170-04
Supplier: Bohlender
Frequently Bought Together