Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Hydroxyurea ≥97% USP

Glass, amber, 500 g
Catalog # SPCMH1181-500GM
Supplier:   Spectrum Chemical
Hydroxyurea ≥97% USP
Catalog # SPCMH1181-500GM
Supplier:  Spectrum Chemical
CAS Number:  127-07-1
Hydroxyurea ≥97% USP

Glass, amber, 500 g

CAS Number:  127-07-1
Price
51.560,00 kr
Log in for availability
1 * 500 g
Product Details & Documents

Hydroxyurea, USP is an antineoplastic drug used in myeloproliferative disorders. It has also been considered useful in treating painful attacks in sickle-cell disease and diseases like AIDS. The USP grade indicates it is graded suitable for personal care, cosmetic and pharmaceutical applications. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Synonyms:  N-(Aminocarbonyl)hydroxylamine, Hydroxycarbamide
  • CAS Number:127-07-1
  • Formula:CH₄N₂O₂
  • MDL Number:MFCD00007943
  • Molecular Weight:76.05 g/mol
  • Merck Index:13,04874
  • Boiling Pt:296 °C (1013 hPa)
  • Melting Pt:142…144 °C
  • Storage Temperature:Ambient
  • EINECS:204-821-7
Specifications

Specification Test Results

  • Assay:
    97% Min.

Specifications

  • Pack type:
    Glass, amber
  • Pk:
    500 g

Specifications