Order Entry
Denmark
ContactUsLinkComponent
Screw In Stirrer Bearings, For screw in into the center neck of BOLA Reactor Lids for centric guidance of stirrer shafts. A special gasket made of PTFE and an FPM o-ring which is compressed by a GL screw cap made of PPS provide a good sealing of the 1 * 1 items
Catalog # BOHLB155-10
Supplier:  Bohlender
undefined
Screw In Stirrer Bearings, For screw in into the center neck of BOLA Reactor Lids for centric guidance of stirrer shafts. A special gasket made of PTFE and an FPM o-ring which is compressed by a GL screw cap made of PPS provide a good sealing of the 1 * 1 items
Catalog # BOHLB155-10
Supplier:  Bohlender
Frequently Bought Together